missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(II) Sulfate Hexahydrate, ≥98%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | 100°C |
Color | Blue Green |
Physical Form | Solid |
Quantity | 5 kg |
Molecular Formula | FeH20N2O14S2 |
Linear Formula | (NH4)2Fe(SO4)2 · 6H2O |
InChI Key | MQLVWQSVRZVNIP-UHFFFAOYSA-L |
SMILES | [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
IUPAC Name | λ²-iron(2+) diammonium hexahydrate disulfate |
Molecular Weight (g/mol) | 392.13 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'