missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodide, 99+%, for analysis, Thermo Scientific Chemicals

Catalog Number p-45518
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
12027-06-4
H4IN
144.94
MFCD00011424
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
25487
CHEBI:63040
azanium;iodide
[NH4+].[I-]
12027-06-4
H4IN
144.94
MFCD00011424
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
25487
CHEBI:63040
azanium;iodide
[NH4+].[I-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 551.0°C
Color White to Yellow
pH 4.5 to 6.5 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula NH4I
Merck Index 15, 526
Solubility Information Solubility in water: 1770g/L (25°C). Other solubilities: 2500g/L (100°C), 270g/L alcohol - 667g/L glycerol, 400g/L methanol
Formula Weight 144.94
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Specific Gravity 2.51
Density 2.5100g/mL
Chemical Name or Material Ammonium iodide, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'