missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodate, 99%, Thermo Scientific Chemicals

Catalog Number p-7049496
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13446-09-8
H4INO3
192.94
MFCD00049684
ZRDJERPXCFOFCP-UHFFFAOYSA-N
ammonium iodate, azanium iodate, iodic acid hio3 , ammonium salt, acmc-1bqbr, ammonium iodate 50g, iodic acid hio3 , ammonium salt 1:1
166805
azanium;iodate
[NH4+].[O-]I(=O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13446-09-8
H4INO3
192.94
MFCD00049684
ZRDJERPXCFOFCP-UHFFFAOYSA-N
ammonium iodate, azanium iodate, iodic acid hio3 , ammonium salt, acmc-1bqbr, ammonium iodate 50g, iodic acid hio3 , ammonium salt 1:1
166805
azanium;iodate
[NH4+].[O-]I(=O)=O

Ammonium iodate is used as an oxidation reagent. Ammonium Iodate-18O3 is the labeled analog of ammonium iodate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150°C
Color White
Physical Form Crystalline
Quantity 50 g
Linear Formula NH4IO3
UN Number UN1479
Solubility Information Soluble in water 20.6g/L @ 20°C
Formula Weight 192.94
Percent Purity 99%
Odor Odorless
Sensitivity Moisture sensitive
Density 3.309 g/mL
Chemical Name or Material Ammonium iodate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'