missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide Solution, ACS Reagent, 28.0 to 30.0% NH3 basis, Honeywell Fluka™
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
2.5 L
6 x 500 mL
12 x 100 mL
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Linear Formula | NH4OH |
Concentration | 28 to 30% NH3 basis |
Beilstein | 3587154 |
Density | 0.9g/mL at 25°C |
Quantity | 25 mL |
IUPAC Name | azanium;hydroxide |
Explosion Limit | 0.27 |
Formula Weight | 35.05g/mol |
Grade | ACS Reagent |
Packaging | Glass Bottle |
Show More |
Safety and Handling
Product Identifier
- Ammonium hydroxide solution
- Danger
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Ammonia, aqueous solution
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'