missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide ACS AR, Macron Fine Chemicals™
Click to view available options
Quantity:
2.5 L
Packaging:
SAFEMOR
Specifications
CAS | 1336-21-6 |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
Quantity | 2.5 L |
SMILES | N.O |
IUPAC Name | amine hydrate |
Molecular Weight (g/mol) | 35.05 |
PubChem CID | 14923 |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'