missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 28.0 to 30.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number 15508654
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
N.O
This item is not returnable. View return policy
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
N.O

High quality chemicals for laboratory and specialized industrial use

Quantity 2.5 L
IUPAC Name azanium;hydroxide
Formula Weight 35.05
Percent Purity 28.0 to 30.0%
Grade BAKER ANALYZED™ ACS Reagent
Packaging Glass Bottle
Specific Gravity 1L = 0.90kg
Chemical Name or Material Ammonium Hydroxide
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'