missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydrogen sulfate, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Ammonium hydrogen sulfate is used as pharmaceutical intermediates and chemical reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 146.9°C |
Physical Form | Lump |
Quantity | 25 g |
UN Number | UN2506 |
Solubility Information | Soluble in water (∽115.1mg/ml at 20°C). Insoluble in acetone,alcohol,and pyridine. |
IUPAC Name | azanium;hydrogen sulfate |
Formula Weight | 115.1 |
Percent Purity | 99.9% |
Sensitivity | Hygroscopic |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'