missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydrogen phosphate, 98%, Thermo Scientific Chemicals
12.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
Description
It is commonly used in fertilizer as plant food as well as a yeast nutrient. Ammonium phosphate dibasic has been used to prepare multi-shell nanoparticles to increase transfection efficiency.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is commonly used in fertilizer as plant food as well as a yeast nutrient. Ammonium phosphate dibasic has been used to prepare multi-shell nanoparticles to increase transfection efficiency.
Solubility
Soluble in w
It is commonly used in fertilizer as plant food as well as a yeast nutrient. Ammonium phosphate dibasic has been used to prepare multi-shell nanoparticles to increase transfection efficiency.
Solubility
Soluble in w
Specifications
Melting Point | 155°C (decomposition) |
CAS | 7783-28-0 |
Quantity | 500 g |
Molecular Formula | H9N2O4P |
Linear Formula | (NH4)2HPO4 |
MDL Number | MFCD00010891 |
Merck Index | 14,542 |
Synonym | diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate |
Solubility Information | Soluble in water |
InChI Key | MNNHAPBLZZVQHP-UHFFFAOYSA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'