missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydrogen Carbonate, Certified AR for analysis, Fisher Chemical™

Catalog Number 10141800
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
1066-33-7
C3H8N2O9Zn2
346.86
12138
QOPSSYZLVIENBR-UHFFFAOYSA-J
dizinc(2+) diammonium tricarbonate
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
This item is not returnable. View return policy
1066-33-7
C3H8N2O9Zn2
346.86
12138
QOPSSYZLVIENBR-UHFFFAOYSA-J
dizinc(2+) diammonium tricarbonate
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Ammonium Hydrogen Carbonate, Certified AR for analysis

TRUSTED_SUSTAINABILITY
Melting Point 105°C
Color White
pH 8
Physical Form Powder/Solid
Quantity 1 kg
Formula Weight 79.06g/mol
Packaging Plastic powder jar
Vapor Pressure 67 hPa at 20°C
compliance-icons
Product Identifier
  • Ammonium bicarbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'