missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hexafluorotitanate, 99.99% (metals basis), Thermo Scientific Chemicals
63.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Ammonium hexafluorotitanate is used as an anti-corrosive cleaning agent. It is also used for the production of ceramics and glass. Further, it is used in the preparation of synthetic gems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Formula Weight | 197.95 |
Percent Purity | 99.99% |
Assay | (metals basis) |
Chemical Name or Material | Ammonium hexafluorotitanate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'