missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hexafluorophosphate, 99.5%, Thermo Scientific Chemicals
36.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Ammonium hexafluorophosphate is used as a preservative. It is used as a starting material in the preparation of other hexafluorophosphates. Further, it is used in synthesis of organometalic compounds, especially rhodicinium hexafluorophosphate, which has inert and non-coordinating counter ion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 198°C |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.5% |
Solubility Information | Freely soluble in water (75g/100ml). Soluble in acetone,methanol,ethanol,methyl acetate.Soluble in water. |
Formula Weight | 163 |
Sensitivity | Hygroscopic |
Density | 2.180 g/mL |
Chemical Name or Material | Ammonium hexafluorophosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'