missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorogermanate, 99.99% min (metals basis), Thermo Scientific Chemicals

Catalog Number p-7058752
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
16962-47-3
H8F6GeN2
MFCD00010880
This item is not returnable. View return policy
16962-47-3
H8F6GeN2
MFCD00010880

Ammonium hexafluorogermanate is used to substitutes Al with Ge in Y type zeolites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 5 g
Linear Formula (NH4)2GeF6
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 222.71
Percent Purity ≥99.99%
Density 2.564 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexafluorogermanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'