missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ammonium hexafluoroaluminate, 99.8% (metals basis)

Preparation of aluminum fluoride

Brand:  Alfa Aesar™ 039128.14

Code : NEW

Additional Details : CAS Number : 7784-19-2 Weight : 0.02500kg Transport : UN number : 3288 Chem class : 6.1 Pack group : III


Quantity 25g

 View more versions of this product

Product Code. 11373508

  • £154.02 / 25g


Estimated Shipment date
from Supplier 29-05-2018
Add to basket


Specifications

Chemical Identifiers

Ammonium hexafluoroaluminate
(NH4)3AlF6
MFCD00042144
14,524
7784-19-2
195.09
UN3288

Specifications

Ammonium hexafluoroaluminate
Powder
(metals basis)
195.09
UN3288
Freely soluble in water
−80 Mesh
White
7784-19-2
99.8%
(NH4)3AlF6
MFCD00042144
14,524
1.78g/mL
Odorless
25g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ammonium hexafluoroaluminate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2

Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H330-Fatal if inhaled.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P320-Specific treatment is urgent (see on this label).
  • P361-Take off immediately all contaminated clothing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS