missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hexachlororuthenate(IV), Ru 28.4% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Ammonium hexachlororuthenate(IV) is used in laboratory chemicals and manufacture of other substances. Ammonium hexachlororuthenate (ACR) is a salt precipitated during a substitution reaction between ammonium and ruthenate dissolved in a strong hydrochloric acid medium. This precipitation reaction is used within the platinum industry as a means of recovering and purifying ruthenium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Red to Brown |
Physical Form | Crystalline Powder |
Quantity | 1 g |
Solubility Information | Soluble in water. |
IUPAC Name | diazanium;hexachlororuthenium(2-) |
Formula Weight | 349.87 |
Concentration or Composition (by Analyte or Components) | Ru 28.4% min. |
Odor | Odorless |
Sensitivity | Hygroscopic |
Chemical Name or Material | Ammonium hexachlororuthenate(IV) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'