missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ammonium hexachlororuthenate(IV), Ru 28.4% min

Brand:  Alfa Aesar™ 011874.06

Code : NEW

Additional Details : CAS Number : 18746-63-9 Weight : 0.00500kg Transport : UN number : 3288 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11301958

  • £197.90 / 5g


Estimated Shipment date
from Supplier 26-04-2018
Add to basket

Specifications

Chemical Identifiers

Ammonium hexachlororuthenate(IV)
18746-63-9
349.87
MFCD00015962

Specifications

Ammonium hexachlororuthenate(IV)
Crystalline Powder
MFCD00015962
Odorless
Red-brown
18746-63-9
349.87
Ru 28.4% min.
Hygroscopic
5g
Videos
SDS