missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachlororhodate(III) hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11321858
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
15336-18-2
Cl6H12N3Rh
369.72
MFCD00003394
QNMMDVLCBKRUJU-UHFFFAOYSA-K
ammonium hexachlororhodate iii, triammonium hexachlororhodate, triammonium hexachlororhodate 3-, epitope id:151523, rhodium iii-ammonium chloride, ammonium hexachloridorhodate 3-, ammonium hexachloridorhodate iii, ammonium hexachlororhodate iii , reagent grade
16211509
CHEBI:60128
triazanium;hexachlororhodium(3-)
[NH4+].[NH4+].[NH4+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl
This item is not returnable. View return policy
15336-18-2
Cl6H12N3Rh
369.72
MFCD00003394
QNMMDVLCBKRUJU-UHFFFAOYSA-K
ammonium hexachlororhodate iii, triammonium hexachlororhodate, triammonium hexachlororhodate 3-, epitope id:151523, rhodium iii-ammonium chloride, ammonium hexachloridorhodate 3-, ammonium hexachloridorhodate iii, ammonium hexachlororhodate iii , reagent grade
16211509
CHEBI:60128
triazanium;hexachlororhodium(3-)
[NH4+].[NH4+].[NH4+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl

Ammonium hexachlororhodate(III) hydrate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color Red
Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 369.74 (Anhydrous)
Percent Purity 99.99%
Grade Premion™
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ammonium hexachlororhodate(III) hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'