missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroplatinate(IV), Pt 43.4% min, Thermo Scientific Chemicals

Catalog Number p-7049093
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16919-58-7
Cl6H8N2Pt
443.86
MFCD00010886
PCCGQTHFYHJATL-UHFFFAOYSA-J
ammonium hexachloroplatinate iv, ammonium chloroplatinate, ammonium hexachloroplatinate, ammonium platinic chloride, ammonium platinum chloride, diammonium hexachloroplatinate 2-, diammonium platinum hexachloride, ammoniumhexachloroplatinate, epitope id:151516, platinum iv-ammonium chloride
16211460
CHEBI:59604
diazanium;hexachloroplatinum(2-)
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
This item is not returnable. View return policy
16919-58-7
Cl6H8N2Pt
443.86
MFCD00010886
PCCGQTHFYHJATL-UHFFFAOYSA-J
ammonium hexachloroplatinate iv, ammonium chloroplatinate, ammonium hexachloroplatinate, ammonium platinic chloride, ammonium platinum chloride, diammonium hexachloroplatinate 2-, diammonium platinum hexachloride, ammoniumhexachloroplatinate, epitope id:151516, platinum iv-ammonium chloride
16211460
CHEBI:59604
diazanium;hexachloroplatinum(2-)
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Electroplating of platinum, manufacturing of platinum sponge

Electroplating of platinum, manufacturing of platinum spongeAmmonium hexachloroplatinate(IV) is used in electroplating of platinum and to make spongy platinum. It reacts with copper(II) acetate and hydrogen sulfide to prepare Cu2PtIIPt3IVS8 which finds application in diamagnetic materials and semiconductors. It is also used in platinum plating. Further, it acts as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color Yellow
Physical Form Powder
Quantity 1 g
UN Number UN3288
Merck Index 14,548
Solubility Information Slightly soluble in water; Insoluble in alcohol,ether and concentrated hydrochloric acid.
Formula Weight 443.89
Concentration or Composition (by Analyte or Components) Pt 43.4% min.
Sensitivity Hygroscopic
Density 3.06 g/mL
Chemical Name or Material Ammonium hexachloroplatinate(IV)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'