missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloropalladate(IV), 99.9% (metals basis), Pd 29%, Thermo Scientific Chemicals

Catalog Number p-7048622
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
19168-23-1
Cl6H8N2Pd
355.20
MFCD00015959
AZUQNARGWZAFRM-UHFFFAOYSA-J
ammonium hexachloropalladate iv, ammonium hexachloropalladate, diammonium hexachloropalladate, diammonium hexachloropalladate 2-, ccris 7829, hexachloropalladate 2-diammonium, ammonium chloropalladate nh4 2pdcl6, palladate 2-, hexachloro-, diammonium, oc-6-11, palladate 2-, hexachloro-, diammonium, epitope id:151517
62732
CHEBI:60124
[NH4+].[NH4+].Cl[Pd++](Cl)(Cl)(Cl)(Cl)Cl
This item is not returnable. View return policy
19168-23-1
Cl6H8N2Pd
355.20
MFCD00015959
AZUQNARGWZAFRM-UHFFFAOYSA-J
ammonium hexachloropalladate iv, ammonium hexachloropalladate, diammonium hexachloropalladate, diammonium hexachloropalladate 2-, ccris 7829, hexachloropalladate 2-diammonium, ammonium chloropalladate nh4 2pdcl6, palladate 2-, hexachloro-, diammonium, oc-6-11, palladate 2-, hexachloro-, diammonium, epitope id:151517
62732
CHEBI:60124
[NH4+].[NH4+].Cl[Pd++](Cl)(Cl)(Cl)(Cl)Cl

Ammonium hexachloropalladate(IV) is used as analytical reagent and as a catalyst in the chemical synthesis. It is also used as raw material and intermediates in organic synthesis and pharmaceutical field. Further, it is used in emission spectrographic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red to Orange
Physical Form Crystalline
Quantity 2 g
UN Number UN3077
Solubility Information Slightly soluble in water.
IUPAC Name diammonium hexachloropalladiumbis(ylium)
Formula Weight 355.2
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 29%
Odor Odorless
Sensitivity Hygroscopic
Density 2.42 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexachloropalladate(IV)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'