missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroosmate(IV), Premion™, 99.99% (metals basis), Os 42.8% min, Thermo Scientific Chemicals

Catalog Number p-7048542
Click to view available options
Quantity:
0.5 g
1 g
5 g
This item is not returnable. View return policy
12125-08-5
Cl6H8N2Os
439.00
MFCD00010883
JRHMYOOMZKAPKT-UHFFFAOYSA-J
ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
11729867
[NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl
This item is not returnable. View return policy
12125-08-5
Cl6H8N2Os
439.00
MFCD00010883
JRHMYOOMZKAPKT-UHFFFAOYSA-J
ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
11729867
[NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl

Ammonium hexachloroosmate(IV) is suitable for glass, optic and ceramic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 170°C (sublimation)
Color Brown to Black
Physical Form Powder
Quantity 0.5 g
Linear Formula (NH4)2OsCl6
Merck Index 14,536
Solubility Information Soluble in water.
IUPAC Name diazanium;hexachloroosmium(2-)
Formula Weight 438.99
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Os 42.8% min.
Sensitivity Hygroscopic
Density 2.9 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexachloroosmate(IV)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'