missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hexachloroiridate(IV), Premion™, 99.994% (metals basis), Ir 41% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.1 g
1 g
5 g
Description
It is a key intermediate in the isolation of iridium from ores.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | Black-Red |
Physical Form | Crystalline Powder |
Quantity | 5 g |
Solubility Information | Soluble in water. |
IUPAC Name | ammonium hexachloroiridate |
Formula Weight | 441.02 |
Percent Purity | 99.994% |
Grade | Premion™ |
Concentration or Composition (by Analyte or Components) | Ir 41% min. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'