missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroiridate(IV), 99% (metals basis), Ir 43% min, Thermo Scientific Chemicals

Catalog Number 11311699
Click to view available options
Quantity:
1 g
5 g
16940-92-4
Cl6H8IrN2
440.99
MFCD00010881
QOPXOJZBUJGPAG-UHFFFAOYSA-J
ammonium hexachloroiridate iv, ammonium chloroiridate, diammonium hexachloroiridate, diammonium hexachloroiridate 2-, ammonium hexachloroiridate, epitope id:151519, iridium iv-ammonium chloride, iridium iii-ammonium chloride, ammonium hexachloridoiridate iv, diazanium;hexachloroiridium 2-
16211476
CHEBI:60132
[NH4+].[NH4+].Cl[Ir+4](Cl)(Cl)(Cl)(Cl)Cl
16940-92-4
Cl6H8IrN2
440.99
MFCD00010881
QOPXOJZBUJGPAG-UHFFFAOYSA-J
ammonium hexachloroiridate iv, ammonium chloroiridate, diammonium hexachloroiridate, diammonium hexachloroiridate 2-, ammonium hexachloroiridate, epitope id:151519, iridium iv-ammonium chloride, iridium iii-ammonium chloride, ammonium hexachloridoiridate iv, diazanium;hexachloroiridium 2-
16211476
CHEBI:60132
[NH4+].[NH4+].Cl[Ir+4](Cl)(Cl)(Cl)(Cl)Cl

Iridium complexes are generated from ammonium hexachloroiridate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color Black-Red
Physical Form Crystalline Powder
Quantity 1 g
Solubility Information Soluble in hydrochloric acid,lightly soluble in water,insoluble in alcohol
IUPAC Name diazanium;hexachloroiridium(2-)
Formula Weight 441.02
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Ir 43% min.
Sensitivity Hygroscopic
Density 2.86 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexachloroiridate(IV)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'