missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroiridate(III) hydrate, Thermo Scientific™

Catalog Number p-7049063
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
29796-57-4
Cl6H14IrN3O
477.049
MFCD00003393
LJHBEBFYNMDGCZ-UHFFFAOYSA-K
ammonium hexachloroiridate iii hydrate, triammonium hexachloroiridiumtriuide hydrate
18366487
triazanium;hexachloroiridium(3-);hydrate
[NH4+].[NH4+].[NH4+].O.Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl
This item is not returnable. View return policy
29796-57-4
Cl6H14IrN3O
477.049
MFCD00003393
LJHBEBFYNMDGCZ-UHFFFAOYSA-K
ammonium hexachloroiridate iii hydrate, triammonium hexachloroiridiumtriuide hydrate
18366487
triazanium;hexachloroiridium(3-);hydrate
[NH4+].[NH4+].[NH4+].O.Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Green
Physical Form Crystalline Powder
Quantity 1 g
Formula Weight 459.06 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Ammonium hexachloroiridate(III) hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'