missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexabromoplatinate(IV), 99.9% (metals basis), Pt 28%, Thermo Scientific™

Catalog Number p-7049055
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
17363-02-9
Br6H8N2Pt
710.59
MFCD00044261
YLDMEZNZYLWNNL-UHFFFAOYSA-J
diazanium;hexabromoplatinum 2-, diammonium hexabromoplatinumdiuide, platinate 2-,hexabromo-,ammonium 1:2 , oc-6-11
16211461
diazanium;hexabromoplatinum(2-)
[NH4+].[NH4+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Pt+4]
This item is not returnable. View return policy
17363-02-9
Br6H8N2Pt
710.59
MFCD00044261
YLDMEZNZYLWNNL-UHFFFAOYSA-J
diazanium;hexabromoplatinum 2-, diammonium hexabromoplatinumdiuide, platinate 2-,hexabromo-,ammonium 1:2 , oc-6-11
16211461
diazanium;hexabromoplatinum(2-)
[NH4+].[NH4+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Pt+4]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 145°C (decomposition)
Physical Form Powder
Quantity 1 g
Formula Weight 710.62
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 28%
Sensitivity Hygroscopic
Density 4.265
Assay (metals basis)
Chemical Name or Material Ammonium hexabromoplatinate(IV)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'