missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexabromoosmate(IV), 99.9% (metals basis), Os 26.5 % min, Thermo Scientific Chemicals

Catalog Number p-7049069
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
24598-62-7
Br6H8N2Os
705.73
MFCD00049389
ZPHFWWAVHILXPL-UHFFFAOYSA-J
diammonium hexabromoosmiumdiuide, osmate 2-,hexabromo-,ammonium 1:2 , oc-6-11
11136267
diazanium;hexabromoosmium(2-)
[NH4+].[NH4+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Os+4]
This item is not returnable. View return policy
24598-62-7
Br6H8N2Os
705.73
MFCD00049389
ZPHFWWAVHILXPL-UHFFFAOYSA-J
diammonium hexabromoosmiumdiuide, osmate 2-,hexabromo-,ammonium 1:2 , oc-6-11
11136267
diazanium;hexabromoosmium(2-)
[NH4+].[NH4+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Os+4]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black
Physical Form Powder
Quantity 1 g
Formula Weight 705.73
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Os 26.5 % min.
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ammonium hexabromoosmate(IV)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'