missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Formate, Optima™ LC/MS Grade, Fisher Chemical™
85.13 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
50g
Description
CH5NO2, CAS Number-540-69-2, 50g, White, 63.06g/mol, 115 deg.C, 13103, 6.0 to 7.0, Solid
Specifications
Melting Point | 115°C |
CAS | 540-69-2 |
Color | White |
pH | 6.0 to 7.0 |
Physical Form | Solid |
Quantity | 50 g |
Molecular Formula | CH5NO2 |
MDL Number | 13103 |
InChI Key | VZTDIZULWFCMLS-UHFFFAOYSA-N |
SMILES | N.OC=O |
Show More |
Safety and Handling
Product Identifier
- Ammonium formate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'