missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 98+%, extra pure, Thermo Scientific Chemicals

Catalog Number p-45471
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Drum
Plastic bottle
12125-01-8
H4FN
37.04
MFCD00011423
12125-01-8
H4FN
37.04
MFCD00011423

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless or White
Physical Form Adhering Crystals and/or Chunks
Quantity 100 g
Assay Percent Range 98+%
Linear Formula NH4F
Merck Index 15, 519
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 37.04
Percent Purity ≥98%
Grade Extra Pure
Packaging Plastic bottle
Residue after Ignition 0.01 % max. (SO4)
Chemical Name or Material Ammonium fluoride, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'