missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 0.5M solution in methanol, ACROS Organics™

Brand:  Acros Organics 393018000

Code : 29

Additional Details : CAS Number : 12125-01-8 Weight : 0.70600kg Transport : UN number : 1992 Chem class : 3 Pack group : II


Quantity 800mL
Packaging Plastic bottle

 View more versions of this product

Product Code. 10445085

  • £284.00 / 800mL


Estimated Shipment
20-06-2018
Add to basket


Specifications

Chemical Identifiers

Ammonium fluoride
H4FN
37.04
67-56-1
NH4F
MFCD00011423

Specifications

Ammonium fluoride
Liquid
NH4F
MFCD00011423
Solubility in water: soluble.
0.8200g/mL
Solubility in water: soluble.
65.0°C
− 98.0°C
11°C
67-56-1
H4FN
37.04
15, 519
0.45 to 0.55M
Plastic bottle
0.82
Clear colorless
800mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ammonium fluoride, 0.5M solution in methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

SDS