missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number p-4228245
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
7722-76-1
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
24402
CHEBI:62982
phosphoric acid amine
N.OP(O)(O)=O
7722-76-1
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
24402
CHEBI:62982
phosphoric acid amine
N.OP(O)(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 190.0°C
Color White or Colorless
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.999%
Linear Formula NH4H2PO4
Merck Index 15,541
Solubility Information Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
Formula Weight 115.03
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Ammonium dihydrogen phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'