Learn More
Ammonium citrate tribasic, 97+%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Ammonium citrate tribasic is widely utilized as a buffering agent and emulsifier in food industry. It acts as a complexing agent in electroplating industry and is also used for metal surface cleaning. It is employed in chemical analysis and in water treatment. It serves as a chemical reagent in analytical chemistry to determine the efficient phosphate levels in phosphate salts and fertilizer. It is used in industries for water treatment, as a ceramic dispersant, a detergent material and a component for soil improvement. It is used in medical and electronics industries as an auxiliary infiltration agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water. Slightly soluble in alcohol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∼180°C (decomposition) |
Assay Percent Range | >97% |
Quantity | 100 g |
Solubility Information | Soluble in water. Slightly soluble in alcohol. |
Formula Weight | 243.22 |
Chemical Name or Material | Ammonium citrate tribasic |
RUO – Research Use Only