missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 340°C (sublimation)(literature) |
CAS Max % | ≤100.0000% |
Quantity | 25 kg |
Assay Percent Range | 99.5 to 100.5% |
Linear Formula | NH4Cl |
Beilstein | 4371014 |
SMILES | [NH4+].[Cl-] |
Formula Weight | 53.49 |
Percent Purity | 99.5 to 100.5% (calc. to the dried substance) |
Grade | puriss. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'