missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog Number p-4407216
Click to view available options
Quantity:
1 kg
5 kg
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
25517
CHEBI:31206
N.Cl
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
25517
CHEBI:31206
N.Cl

Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacturing of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 340°C (Sublimation)
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99.5% min
Solubility Information Soluble in liquid ammonia,acetone,hydrazine and alcohol. Insoluble in diethyl ether and ethyl acetate.
IUPAC Name azanium;chloride
Formula Weight 53.49
Grade ACS Reagent
Refractive Index 1.642
Sensitivity Hygroscopic
Density 1.53
Chemical Name or Material Ammonium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'