missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, 99+%, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072811
30.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


45.44 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
N.Cl
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
N.Cl
Melting Point 340°C
Color White
pH 5
Physical Form Solid
Quantity 500 g
Formula Weight 53.49g/mol
Packaging Plastic powder jar
Vapor Pressure 1.3mbar 160
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'