missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, 99.5%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3734638
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic bottle
Plastic drum
This item is not returnable. View return policy
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
25517
CHEBI:31206
This item is not returnable. View return policy
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
25517
CHEBI:31206

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 340.0°C
Color White
pH 4.5 to 5.5 (5% soln. at 20°C)
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5% min. (Argentometry)
Linear Formula NH3·HCl
Fieser 15,13
Merck Index 15, 506
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, practically insoluble in acetone, ether and, ethyl acetate
SMILES N.Cl
IUPAC Name amine hydrochloride
Formula Weight 53.49
Percent Purity 99.5%
Grade Extra Pure
Packaging Plastic bottle
Density 1.5200g/mL
Chemical Name or Material Ammonium chloride
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'