missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium cerium(IV) sulfate dihydrate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | Ce(NH4)4(SO4)4 · 2H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 632.55g/mol |
Percent Purity | ≥94% |
Chemical Name or Material | Ammonium cerium(IV) sulfate dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'