missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cerium(IV) nitrate, 99%, Reagent ACS, Acros Organics™

Brand:  Acros Organics 423545000

Code : 29

Additional Details : CAS Number : 16774-21-3 Weight : 0.55000kg Transport : UN number : 3085 Chem class : 5.1 Pack group : II


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10483551

  • £153.30 / 500g


EU Stock 8
Add to basket


Description

Description

Oxidation reagent
Specifications

Chemical Identifiers

Ammonium cerium(IV) nitrate
16774-21-3
99%
H8CeN8O18
548.22
Ceric ammonium nitrate, CAN
98.5%
ACS Reagent Grade
99%
(NH4)2Ce(NO3)6
MFCD00151121

Specifications

Ammonium cerium(IV) nitrate
16774-21-3
Powder or Flakes
99%
(NH4)2Ce(NO3)6
MFCD00151121
Ceric ammonium nitrate, CAN
100ppm max.
0.05 % max. (in dilute H2SO4)
Glass bottle
Solubility in water: 1410g/L (20°C). Other solubilities: insoluble in fat, soluble in alcohol
107.0° to 108.0°C
98.5%
ACS Reagent Grade
99%
H8CeN8O18
548.22
01,120; 02,63; 03,44; 04,71; 05,101; 06,99; 09,99; 10,79; 15,70; 16,66; 17,68
100ppm max.
1.05
50ppm max.
200ppm max.
White to pale yellow-beige
500g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ammonium cerium(IV) nitrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P221-Take any precaution to avoid mixing with combustibles .
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

SDS