missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Cerium(IV) Nitrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
100 g
250 g
Specifications
Melting Point | 107°C |
Color | Orange |
pH | 1 |
Physical Form | Powder/Solid |
Quantity | 250 g |
Formula Weight | 548.22g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'