missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, Extra Pure, Powder, SLR, Fisher Chemical™

Catalog Number p-7072807
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
506-87-6
CH8N2O3
96.09
10890
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
carbonic acid diamine
N.N.OC(O)=O
This item is not returnable. View return policy
506-87-6
CH8N2O3
96.09
10890
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
carbonic acid diamine
N.N.OC(O)=O
Melting Point 58°C
Color White
pH 9.4
Physical Form Solid
Quantity 1 kg
Formula Weight 96.09g/mol
Packaging Plastic powder jar
Vapor Pressure 69 hPa at 20°C
compliance-icons
Product Identifier
  • Ammonium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'