missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium bromide, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 235.0°C |
Melting Point | 452.0°C |
Quantity | 2.5 kg |
Linear Formula | NH4Br |
Merck Index | 15,502 |
Solubility Information | Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C), freely soluble in alcohol and acetone, slightly soluble in ether, practically insoluble in ethyl acetate |
Formula Weight | 97.94 |
Grade | ACS Reagent |
Packaging | Plastic Bottle |
Chemical Name or Material | Ammonium Bromide |
Safety and Handling
Product Identifier
- Ammonium bromide
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'