missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bromide, 99+%, for analysis, Thermo Scientific Chemicals

Catalog Number p-4509821
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
12124-97-9
BrH4N
97.94
MFCD00011419
SWLVFNYSXGMGBS-UHFFFAOYSA-N
ammonium bromide
[NH4+].[Br-]
This item is not returnable. View return policy
12124-97-9
BrH4N
97.94
MFCD00011419
SWLVFNYSXGMGBS-UHFFFAOYSA-N
ammonium bromide
[NH4+].[Br-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 235.0°C
Melting Point 452.0°C
Color White
pH 4.5 to 6.0 (5% soln.)
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99% min. (on dry substance) (Argentometry)
Linear Formula NH4Br
Merck Index 15,502
Solubility Information Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C), freely soluble in alcohol and acetone, slightly soluble in ether, practically insoluble in ethyl acetate
Formula Weight 97.94
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.2% (105°C) max.
Packaging Glass bottle
Chemical Name or Material Ammonium bromide
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'