missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ammonium bis(oxalato)palladium(II) dihydrate, Premion™, 99.99% (metals basis)

Brand:  Alfa Aesar™ 043475.06

Code : NEW

Additional Details : Weight : 0.00500kg Transport : UN number : 3288 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11315797

  • £226.52 / 5g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Ammonium bis (oxalato)palladium (II) dihydrate
(NH4)2Pd(C2O4)2·2H2O
MFCD02684521
Premion™
354.57 (318.54 Anhydrous)
UN3288

Specifications

Ammonium bis (oxalato)palladium (II) dihydrate
Crystalline
(metals basis)
354.57 (318.54 Anhydrous)
UN3288
Premion™
99.99%
(NH4)2Pd(C2O4)2·2H2O
MFCD02684521
5g
Videos
SDS