missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Bifluoride, Flake, BAKER ANALYZED™ Reagent, J.T. Baker™
474.05 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Flakes |
Quantity | 2.5 kg |
Formula Weight | 57.04 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.51kg |
Chemical Name or Material | Ammonium Bifluoride |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'