missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Bicarbonate, Reagent Grade, ≥99%, Honeywell™
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | 60°C |
Color | Colorless |
pH | 8 (at 20°C) |
Quantity | 1 kg |
Linear Formula | NH4HCO3 |
Beilstein | 4329606 |
IUPAC Name | dizinc(2+) diammonium tricarbonate |
Formula Weight | 79.06g/mol |
Percent Purity | ≥99% |
Odor | Ammoniacal |
Show More |
Safety and Handling
Product Identifier
- Ammonium bicarbonate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'