missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate (White Crystals), Fisher BioReagents

Catalog Number p-24395
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
This item is not returnable. View return policy
631-61-8
C2H7NO2
77.083
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]
This item is not returnable. View return policy
631-61-8
C2H7NO2
77.083
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]
Ammonium Acetate can be used to precipitate DNA from enzymatic reactions.
TRUSTED_SUSTAINABILITY
Melting Point 114°C
Color White
pH 7.0
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥97 %
Linear Formula CH3COONH4
Merck Index 15, 492
Formula Weight 77.08
Percent Purity ≥97%
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Ammonium Acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'