missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amiodarone hydrochloride, ACROS Organics™
£15.07 - £123.05
Chemical Identifiers
CAS | 19774-82-4 |
---|---|
Molecular Formula | C25H29I2NO3·HCl |
Molecular Weight (g/mol) | 681.77 |
InChI Key | ITPDYQOUSLNIHG-UHFFFAOYSA-N |
Synonym | amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron |
PubChem CID | 441325 |
IUPAC Name | (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride |
SMILES | CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15592223
|
Acros Organics
457980010 |
1g |
£15.07
1g |
Estimated Shipment 22-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15502233
|
Acros Organics
457980050 |
5g |
£31.47
5g |
Estimated Shipment 22-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15512233
|
Acros Organics
457980250 |
25g |
£123.05
25g |
Estimated Shipment 22-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
19774-82-4 | |
681.77 | |
amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride |
C25H29I2NO3·HCl | |
ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
441325 | |
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl |
Specifications
Amiodarone hydrochloride | |
0.5% max. | |
156.0°C | |
19774-82-4 | |
amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron | |
ITPDYQOUSLNIHG-UHFFFAOYSA-N | |
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride | |
441325 | |
≥97.5% (HPLC) |
Authentic | |
0.1% max. | |
1g | |
C25H29I2NO3·HCl | |
Solubility in water: insoluble. | |
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl | |
681.77 | |
681.77 |