missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amiodarone hydrochloride, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Melting Point | 156.0°C |
Loss on Drying | 0.5% max. |
Infrared Spectrum | Authentic |
Quantity | 25 g |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 681.77 |
Percent Purity | ≥97.5% (HPLC) |
Chemical Name or Material | Amiodarone hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'