missing translation for 'onlineSavingsMsg'
Learn More

Amiodarone hydrochloride, Thermo Scientific™

Catalog Number 15502233
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
19774-82-4
C25H29I2NO3·HCl
681.77
ITPDYQOUSLNIHG-UHFFFAOYSA-N
amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
441325
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl
This item is not returnable. View return policy
19774-82-4
C25H29I2NO3·HCl
681.77
ITPDYQOUSLNIHG-UHFFFAOYSA-N
amiodarone hydrochloride, amiodarone hcl, amiodar, nexterone, pacerone, amiodaronum hydrochloride, ritmocardyl, rythmarone, angoron
441325
(2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride
CCCCC1=C(C2=CC=CC=C2O1)C(=O)C3=CC(=C(C(=C3)I)OCCN(CC)CC)I.Cl
Melting Point 156.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 681.77
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Amiodarone hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'