missing translation for 'onlineSavingsMsg'
Learn More

Aminomethanesulfonic acid, 97%, ACROS Organics™

£18.69 - £96.94

Chemical Identifiers

CAS 13881-91-9
Molecular Formula CH5NO3S
Molecular Weight (g/mol) 111.12
MDL Number MFCD00008124
InChI Key OBESRABRARNZJB-UHFFFAOYSA-N
Synonym methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid
PubChem CID 83791
IUPAC Name aminomethanesulfonic acid
SMILES C(N)S(=O)(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10041800
SDS View Documents
Acros Organics
155740500
50g Glass bottle
£18.69
50g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
13438378
View Documents
Acros Organics
155745000
500g Glass bottle
£96.94
500g
View alternative products
Save Up to 
»
In stock, ready to ship: 2 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

13881-91-9
111.12
OBESRABRARNZJB-UHFFFAOYSA-N
83791
C(N)S(=O)(=O)O
CH5NO3S
MFCD00008124
methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid
aminomethanesulfonic acid

Specifications

Aminomethanesulfonic acid
Glass bottle
180.0°C to 185.0°C
97%
97%
H2NCH2SO3H
01,583
OBESRABRARNZJB-UHFFFAOYSA-N
aminomethanesulfonic acid
83791
Fine Powder
Authentic
White
50g
13881-91-9
CH5NO3S
MFCD00008124
methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid
C(N)S(=O)(=O)O
111.12
111.12
97%
Safety and Handling

Safety and Handling

Product Identifier
  • Aminomethanesulfonic acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS