missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine nitrate, 98%, cont. ca 10% water, Thermo Scientific Chemicals

Catalog Number p-7045697
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
10308-82-4
CH7N5O3
137.10
MFCD00013174
PMGFHEJUUBDCLU-UHFFFAOYSA-N
aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acid
165859
2-aminoguanidine;nitric acid
O[N+]([O-])=O.NN=C(N)N
This item is not returnable. View return policy
10308-82-4
CH7N5O3
137.10
MFCD00013174
PMGFHEJUUBDCLU-UHFFFAOYSA-N
aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acid
165859
2-aminoguanidine;nitric acid
O[N+]([O-])=O.NN=C(N)N

Aminoguanidine nitrate is used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoguanidine nitrate is used as a pharmaceutical raw material.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with reducing agents, strong oxidizing agents, organic materials and metal powders.
TRUSTED_SUSTAINABILITY
Melting Point 138°C to 142°C
Quantity 25 g
UN Number UN1479
Beilstein 3916056
Solubility Information Soluble in water.
Formula Weight 137.1
Percent Purity 98%
Assay cont. ≈10% water
Chemical Name or Material Aminoguanidine nitrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'