missing translation for 'onlineSavingsMsg'
Learn More

(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers, Thermo Scientific Chemicals

Catalog Number p-7059082
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
74113-77-2
H2NC2H4NHCH2C6H4C2H4Si(OCH3)3
MFCD00054775
aminoethy-laminomethyl phenethyl-trimethoxysilane, 4-2-aminoethylaminomethyl phenethyltrimethoxysilane, n1-4-2-trimethoxysilyl ethyl benzyl ethane-1,2-diamine, n'-4-2-trimethoxysilylethyl phenyl methyl ethane-1,2-diamine, 2-aminoethyl 4-2-trimethoxysilyl ethyl phenyl methyl amine
This item is not returnable. View return policy
74113-77-2
H2NC2H4NHCH2C6H4C2H4Si(OCH3)3
MFCD00054775
aminoethy-laminomethyl phenethyl-trimethoxysilane, 4-2-aminoethylaminomethyl phenethyltrimethoxysilane, n1-4-2-trimethoxysilyl ethyl benzyl ethane-1,2-diamine, n'-4-2-trimethoxysilylethyl phenyl methyl ethane-1,2-diamine, 2-aminoethyl 4-2-trimethoxysilyl ethyl phenyl methyl amine

As a crosslinking agent

As a crosslinking agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 128°C (0.2mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 2 g
UN Number UN2735
Flash Point >110°C (230°F)
Formula Weight 298.46
Sensitivity Air and moisture sensitive
Density 1.02 g/mL
Chemical Name or Material (Aminoethylaminomethyl)phenethyltrimethoxysilane, Mixture of m and p isomers

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'