missing translation for 'onlineSavingsMsg'
Learn More

Aminodiphenylmethane, 97%, ACROS Organics™

Brand:  Acros Organics 304320010

Code : 29

Additional Details : CAS Number : 91-00-9 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10676481

  • £8.90 / 1g


Estimated Shipment
09-02-2018
Add to basket

Specifications

Chemical Identifiers

Aminodiphenylmethane
97%
(C6H5)2CHNH2
MFCD00008059
97%
C13H13N
183.25

Specifications

Aminodiphenylmethane
Authentic
1.5950 to 1.5970
Clear colorless to yellow
295.0°C
97%
97%
C13H13N
183.25
12, 1323
Benzhydrylamine
91-00-9
Glass bottle
1.0600g/mL
12.0°C
Liquid
1g
>110°C
(C6H5)2CHNH2
MFCD00008059
15, 1078
Videos
SDS